siRNA Details
| Gene Symbol: Dpt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105965.1 | NP_001099435.1 | 2 | 414 | ||
| XM_006250153.1 | XP_006250215.1 | 2 | 651 | |||
| XM_006250154.1 | XP_006250216.1 | 2 | 651 | |||
| XM_008769624.1 | XP_008767846.1 | 2 | 651 | |||
| GenBank mRNA | BC167054.1 | - | 2 | 409 | 7113939 | |

Dpt
Genomic Map