siRNA Details
| Gene Symbol: Mpc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077643.1 | NP_001071111.1 | 4 | 289 | ||
| XM_006250126.2 | XP_006250188.1 | 4 | 1084 | |||
| GenBank mRNA | FQ215073.1 | - | 4 | 430 | ||
| FQ217960.1 | - | 4 | 358 | |||
| FQ222522.1 | - | 4 | 424 | |||
| FQ224252.1 | - | 4 | 321 | |||

Mpc2
Genomic Map