siRNA Details
| Gene Symbol: TRPC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130698.1 | NP_001124170.1 | 10, 11 | 2621 | ||
| NM_003305.2 | NP_003296.1 | 9, 10 | 2403 | |||
| XM_011532217.2 | XP_011530519.1 | 11, 12 | 2975 | |||
| GenBank mRNA | AB255423.1 | - | 8, 9 | 2163 | ||
| AB255424.1 | - | 9, 10 | 2328 | |||
| AY865574.1 | - | 10, 11 | 2621 | |||
| BC093682.1 | - | 9, 10 | 2403 | 7939527 | ||
| BC093684.1 | - | 9, 10 | 2403 | 7939529 | ||
| Y13758.1 | - | 9, 10 | 2752 | |||

TRPC3
Genomic Map