siRNA Details
| Gene Symbol: Fcgr2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175756.1 | NP_786932.1 | 5 | 814 | ||
| XM_006250235.1 | XP_006250297.1 | 5 | 828 | |||
| GenBank mRNA | BC091113.1 | - | 5 | 808 | 7323350 | |
| X73371.1 | - | 5 | 848 | |||

Fcgr2b
Genomic Map