siRNA Details
| Gene Symbol: Ndufs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011907.1 | NP_001011907.1 | 11 | 1269 | ||
| XM_003751277.2 | XP_003751325.1 | 11 | 1282 | |||
| GenBank mRNA | BC082067.1 | - | 11 | 1269 | 7132777 | |
| FQ215922.1 | - | 11 | 1273 | |||

Ndufs2
Genomic Map