siRNA Details
| Gene Symbol: Ppox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105968.1 | NP_001099438.1 | 11 | 1528 | ||
| XM_006250240.2 | XP_006250302.1 | 10 | 2429 | |||
| XM_006250241.1 | XP_006250303.1 | 11 | 1593 | |||
| XM_006250243.1 | XP_006250305.1 | 10 | 1236 | |||

Ppox
Genomic Map