siRNA Details
Gene Symbol: Ly9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191673.1 | NP_001178602.1 | 3 | 593 | ||
XM_008769732.1 | XP_008767954.1 | 3 | 673 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ly9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191673.1 | NP_001178602.1 | 3 | 593 | ||
XM_008769732.1 | XP_008767954.1 | 3 | 673 |