siRNA Details
| Gene Symbol: Ly9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191673.1 | NP_001178602.1 | 8 | 1545 | ||
| XM_008769732.1 | XP_008767954.1 | 8 | 1631 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ly9| Gene Symbol: Ly9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191673.1 | NP_001178602.1 | 8 | 1545 | ||
| XM_008769732.1 | XP_008767954.1 | 8 | 1631 | |||