siRNA Details
| Gene Symbol: Vangl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105969.1 | NP_001099439.1 | 5 | 944 | ||
| XM_006250276.2 | XP_006250338.1 | 6 | 1070 | |||
| XM_006250277.2 | XP_006250339.1 | 4 | 859 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vangl2| Gene Symbol: Vangl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105969.1 | NP_001099439.1 | 5 | 944 | ||
| XM_006250276.2 | XP_006250338.1 | 6 | 1070 | |||
| XM_006250277.2 | XP_006250339.1 | 4 | 859 | |||