siRNA Details
| Gene Symbol: Ncstn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174864.3 | NP_777353.1 | 4, 5 | 528 | ||
| XM_006250283.1 | XP_006250345.1 | 3, 4 | 383 | |||
| XM_008769733.1 | XP_008767955.1 | 4, 5 | 513 | |||
| XM_008769734.1 | XP_008767956.1 | 4, 5 | 470 | |||
| XM_008769735.1 | XP_008767957.1 | 2, 3 | 231 | |||
| GenBank mRNA | AF510722.1 | - | 3, 4 | 312 | ||
| AY101378.1 | - | 4, 5 | 436 | |||
| BC074006.1 | - | 5, 6 | 643 | |||
| BC167747.1 | - | 3, 4 | 383 | 8365089 | ||

Ncstn
Genomic Map