siRNA Details
| Gene Symbol: Olr1579 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000078.1 | NP_001000078.1 | 1 | 393 | ||
| Gene Symbol: Olr1581 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000533.1 | NP_001000533.1 | 1 | 393 | ||

Olr1579
Genomic Map