siRNA Details
| Gene Symbol: LOC100360457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769811.1 | XP_008768033.1 | 11 | 3696 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100360457| Gene Symbol: LOC100360457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769811.1 | XP_008768033.1 | 11 | 3696 | ||