siRNA Details
| Gene Symbol: Pld5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191674.1 | NP_001178603.1 | 9, 10 | 1351 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pld5| Gene Symbol: Pld5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191674.1 | NP_001178603.1 | 9, 10 | 1351 | ||