siRNA Details
| Gene Symbol: Desi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013873.1 | NP_001013895.1 | 3 | 594 | ||
| XM_006250316.2 | XP_006250378.1 | 3 | 944 | |||
| XM_006250317.2 | XP_006250379.1 | 4 | 1358 | |||
| XM_008769783.1 | XP_008768005.1 | 3 | 901 | |||
| XM_008769784.1 | XP_008768006.1 | 4 | 1077 | |||
| XM_008769785.1 | XP_008768007.1 | 3 | 1205 | |||
| GenBank mRNA | BC083584.1 | - | 3 | 594 | 7131107 | |

Desi2
Genomic Map