siRNA Details
| Gene Symbol: Acbd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182843.1 | NP_878263.1 | 5 | 915 | ||
| GenBank mRNA | AY336075.1 | - | 5 | 915 | ||
| BC083877.1 | - | 5 | 876 | 7130229 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acbd3| Gene Symbol: Acbd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182843.1 | NP_878263.1 | 5 | 915 | ||
| GenBank mRNA | AY336075.1 | - | 5 | 915 | ||
| BC083877.1 | - | 5 | 876 | 7130229 | ||