siRNA Details
| Gene Symbol: Sde2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024970.2 | NP_001020141.2 | 6 | 1133 | ||
| GenBank mRNA | BC091401.1 | - | 3 | 1016 | 7324146 | |
| CV077309.1 | - | 1 | 265 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sde2| Gene Symbol: Sde2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024970.2 | NP_001020141.2 | 6 | 1133 | ||
| GenBank mRNA | BC091401.1 | - | 3 | 1016 | 7324146 | |
| CV077309.1 | - | 1 | 265 | |||