siRNA Details
| Gene Symbol: Tmem63a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134496.2 | NP_001127968.2 | 8 | 750 | ||
| XM_006250364.2 | XP_006250426.1 | 8 | 927 | |||
| XM_006250365.2 | XP_006250427.1 | 8 | 782 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem63a| Gene Symbol: Tmem63a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134496.2 | NP_001127968.2 | 8 | 750 | ||
| XM_006250364.2 | XP_006250426.1 | 8 | 927 | |||
| XM_006250365.2 | XP_006250427.1 | 8 | 782 | |||