siRNA Details
| Gene Symbol: Eprs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024238.1 | NP_001019409.1 | 23 | 3284 | ||
| XM_006250425.2 | XP_006250487.1 | 24 | 3537 | |||
| XM_006250426.2 | XP_006250488.1 | 23 | 3516 | |||
| XM_006250427.2 | XP_006250489.1 | 20 | 2917 | |||
| GenBank mRNA | AY383705.1 | - | 23 | 3284 | ||
| BC088324.1 | - | 3 | 336 | |||

Eprs
Genomic Map