siRNA Details
| Gene Symbol: Slc30a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105985.1 | NP_001099455.1 | 3 | 1068 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc30a10| Gene Symbol: Slc30a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105985.1 | NP_001099455.1 | 3 | 1068 | ||