siRNA Details
| Gene Symbol: Kif21b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105990.1 | NP_001099460.1 | 30 | 4347 | ||
| XM_006249853.2 | XP_006249915.1 | 30 | 4394 | |||
| XM_006249854.2 | XP_006249916.1 | 29 | 4355 | |||
| XM_006249855.2 | XP_006249917.1 | 29 | 4352 | |||
| XM_006249856.2 | XP_006249918.1 | 30 | 4394 | |||
| XM_006249857.2 | XP_006249919.1 | 30 | 4391 | |||
| XM_006249858.2 | XP_006249920.1 | 29 | 4351 | |||

Kif21b
Genomic Map