siRNA Details
| Gene Symbol: RGD1311892 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134502.1 | NP_001127974.1 | 4 | 728 | ||
| XM_006249920.2 | XP_006249982.1 | 4 | 589 | |||
| XM_006249922.2 | XP_006249984.1 | 4 | 875 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1311892| Gene Symbol: RGD1311892 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134502.1 | NP_001127974.1 | 4 | 728 | ||
| XM_006249920.2 | XP_006249982.1 | 4 | 589 | |||
| XM_006249922.2 | XP_006249984.1 | 4 | 875 | |||