siRNA Details
| Gene Symbol: Ddx59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005535.2 | NP_001005535.1 | 7 | 1690 | ||
| NM_001190820.1 | NP_001177749.1 | 8 | 1780 | |||
| GenBank mRNA | BC081871.1 | - | 7 | 1644 | 7106377 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddx59| Gene Symbol: Ddx59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005535.2 | NP_001005535.1 | 7 | 1690 | ||
| NM_001190820.1 | NP_001177749.1 | 8 | 1780 | |||
| GenBank mRNA | BC081871.1 | - | 7 | 1644 | 7106377 | |