siRNA Details
| Gene Symbol: Ddx59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005535.2 | NP_001005535.1 | 2, 3 | 938 | ||
| NM_001190820.1 | NP_001177749.1 | 2, 3 | 938 | |||
| GenBank mRNA | BC081871.1 | - | 2, 3 | 892 | 7106377 | |
| CO398251.1 | - | 1, 2 | 286 | |||

Ddx59
Genomic Map