siRNA Details
| Gene Symbol: Nuak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007617.1 | NP_001007618.1 | 7 | 1197 | ||
| XM_008769444.1 | XP_008767666.1 | 7 | 1898 | |||
| GenBank mRNA | BC081899.1 | - | 7 | 1197 | 7110518 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nuak2| Gene Symbol: Nuak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007617.1 | NP_001007618.1 | 7 | 1197 | ||
| XM_008769444.1 | XP_008767666.1 | 7 | 1898 | |||
| GenBank mRNA | BC081899.1 | - | 7 | 1197 | 7110518 | |