siRNA Details
| Gene Symbol: Xpr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105992.1 | NP_001099462.1 | 5 | 567 | ||
| XM_008769626.1 | XP_008767848.1 | 5 | 1456 | |||
| GenBank mRNA | BC161992.1 | - | 5 | 657 | 9033232 | |
More Information
Set Membership: |
|

Xpr1
Genomic Map