siRNA Details
| Gene Symbol: Glmn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105993.1 | NP_001099463.1 | 11 | 1169 | ||
| XM_008769921.1 | XP_008768143.1 | 10 | 1776 | |||
| XM_008769922.1 | XP_008768144.1 | 11 | 1170 | |||
| XM_008769923.1 | XP_008768145.1 | 11 | 1249 | |||
| XM_008769924.1 | XP_008768146.1 | 10 | 1776 | |||
| XM_008769925.1 | XP_008768147.1 | 9 | 1835 | |||
| XM_008769926.1 | XP_008768148.1 | 9 | 962 | |||
| GenBank mRNA | BC083633.1 | - | 10 | 1109 | ||
| BC100158.1 | - | 11 | 1282 | |||
| BC162027.1 | - | 11 | 1145 | |||

Glmn
Genomic Map