siRNA Details
| Gene Symbol: Ephx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105994.1 | NP_001099464.1 | 6 | 782 | ||
| XM_006250541.1 | XP_006250603.1 | 6 | 835 | |||
| XM_008769927.1 | XP_008768149.1 | 6 | 1694 | |||
| GenBank mRNA | FQ212126.1 | - | 6 | 806 | ||

Ephx4
Genomic Map