siRNA Details
| Gene Symbol: Ephx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105994.1 | NP_001099464.1 | 6 | 738 | ||
| XM_006250541.1 | XP_006250603.1 | 6 | 791 | |||
| XM_008769927.1 | XP_008768149.1 | 6 | 1650 | |||
| GenBank mRNA | FQ212126.1 | - | 6 | 762 | ||
| FQ213206.1 | - | 6 | 664 | |||

Ephx4
Genomic Map