siRNA Details
Gene Symbol: Lrrc8c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037179.1 | NP_001032256.1 | 3, 4 | 408 | ||
XM_006250542.2 | XP_006250604.1 | 1, 2 | 141 | |||
GenBank mRNA | BC100076.1 | - | 3, 4 | 408 | 7386007 |
