siRNA Details
| Gene Symbol: Hsd17b11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004209.1 | NP_001004209.1 | 4 | 545 | ||
| XM_006250627.2 | XP_006250689.1 | 5 | 732 | |||
| XM_006250629.2 | XP_006250691.1 | 5 | 656 | |||
| GenBank mRNA | BC078929.1 | - | 4 | 545 | 7107084 | |
| FQ210820.1 | - | 4 | 561 | |||
| FQ211398.1 | - | 4 | 568 | |||
| FQ222122.1 | - | 4 | 558 | |||

Hsd17b11
Genomic Map