siRNA Details
| Gene Symbol: Gk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004077.1 | NP_001004077.1 | 1 | 259 | ||
| GenBank mRNA | BC079449.1 | - | 1 | 259 | 7115796 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gk2| Gene Symbol: Gk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004077.1 | NP_001004077.1 | 1 | 259 | ||
| GenBank mRNA | BC079449.1 | - | 1 | 259 | 7115796 | |