siRNA Details
| Gene Symbol: Fras1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191595.1 | NP_001178524.1 | 53 | 8715 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fras1| Gene Symbol: Fras1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191595.1 | NP_001178524.1 | 53 | 8715 | ||