siRNA Details
| Gene Symbol: Sdad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006958.1 | NP_001006959.1 | 18 | 1624 | ||
| XM_006250722.2 | XP_006250784.1 | 18 | 1694 | |||
| GenBank mRNA | BC083620.1 | - | 18 | 1624 | 7133993 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sdad1| Gene Symbol: Sdad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006958.1 | NP_001006959.1 | 18 | 1624 | ||
| XM_006250722.2 | XP_006250784.1 | 18 | 1694 | |||
| GenBank mRNA | BC083620.1 | - | 18 | 1624 | 7133993 | |