siRNA Details
| Gene Symbol: Epgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_223280.6 | XP_223280.3 | 4 | 539 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Epgn| Gene Symbol: Epgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_223280.6 | XP_223280.3 | 4 | 539 | ||