siRNA Details
| Gene Symbol: Ankrd17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250763.2 | XP_006250825.1 | 28 | 5520 | ||
| XM_006250764.2 | XP_006250826.1 | 28 | 5517 | |||
| XM_006250765.2 | XP_006250827.1 | 28 | 5466 | |||
| XM_006250766.2 | XP_006250828.1 | 28 | 6102 | |||
| XM_006250767.2 | XP_006250829.1 | 28 | 6402 | |||
| XM_006250768.2 | XP_006250830.1 | 27 | 4692 | |||
| XM_006250769.2 | XP_006250831.1 | 27 | 4689 | |||
| XM_006250770.2 | XP_006250832.1 | 27 | 4638 | |||
| XM_008770011.1 | XP_008768233.1 | 28 | 6099 | |||

Ankrd17
Genomic Map