siRNA Details
| Gene Symbol: Cox18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106000.1 | NP_001099470.1 | 5 | 806 | ||
| XM_006250771.2 | XP_006250833.1 | 5 | 4408 | |||
| XM_006250772.2 | XP_006250834.1 | 4 | 4284 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cox18| Gene Symbol: Cox18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106000.1 | NP_001099470.1 | 5 | 806 | ||
| XM_006250771.2 | XP_006250833.1 | 5 | 4408 | |||
| XM_006250772.2 | XP_006250834.1 | 4 | 4284 | |||