siRNA Details
Gene Symbol: Enam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106001.1 | NP_001099471.1 | 10 | 3527 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Enam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106001.1 | NP_001099471.1 | 10 | 3527 |