siRNA Details
| Gene Symbol: RGD1311575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250863.2 | XP_006250925.1 | 7 | 3642 | ||
| XM_008770114.1 | XP_008768336.1 | 7 | 3651 | |||
| XM_008770115.1 | XP_008768337.1 | 8 | 4809 | |||
| XM_008770116.1 | XP_008768338.1 | 3984 | ||||
| XM_008770117.1 | XP_008768339.1 | 8 | 3980 | |||
| XM_008770118.1 | XP_008768340.1 | 7 | 3673 | |||
| GenBank mRNA | FQ211842.1 | - | 1 | 93 | ||

RGD1311575
Genomic Map