siRNA Details
| Gene Symbol: Lrrc66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008763.1 | NP_001008763.1 | 4 | 1862 | ||
| XM_008770138.1 | XP_008768360.1 | 5 | 2864 | |||
| GenBank mRNA | AY383699.1 | - | 4 | 1862 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc66| Gene Symbol: Lrrc66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008763.1 | NP_001008763.1 | 4 | 1862 | ||
| XM_008770138.1 | XP_008768360.1 | 5 | 2864 | |||
| GenBank mRNA | AY383699.1 | - | 4 | 1862 | ||