siRNA Details
| Gene Symbol: Nfxl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250896.2 | 17 | 2146 | |||
| GenBank mRNA | BC088853.1 | - | 4 | 343 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nfxl1| Gene Symbol: Nfxl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250896.2 | 17 | 2146 | |||
| GenBank mRNA | BC088853.1 | - | 4 | 343 | ||