siRNA Details
| Gene Symbol: Corin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182473.1 | NP_872279.1 | 20 | 2919 | ||
| XM_008770139.1 | XP_008768361.1 | 20 | 2966 | |||
| GenBank mRNA | AY251285.1 | - | 20 | 2919 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Corin| Gene Symbol: Corin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182473.1 | NP_872279.1 | 20 | 2919 | ||
| XM_008770139.1 | XP_008768361.1 | 20 | 2966 | |||
| GenBank mRNA | AY251285.1 | - | 20 | 2919 | ||