siRNA Details
| Gene Symbol: Commd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106004.2 | NP_001099474.1 | 2 | 147 | ||
| GenBank mRNA | BC159414.1 | - | 2 | 147 | 8368074 | |
| FQ212906.1 | - | 2 | 156 | |||
| FQ221260.1 | - | 2 | 148 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Commd8| Gene Symbol: Commd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106004.2 | NP_001099474.1 | 2 | 147 | ||
| GenBank mRNA | BC159414.1 | - | 2 | 147 | 8368074 | |
| FQ212906.1 | - | 2 | 156 | |||
| FQ221260.1 | - | 2 | 148 | |||