siRNA Details
| Gene Symbol: Gabra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135779.1 | NP_001129251.1 | 9 | 1050 | ||
| XM_006250931.2 | XP_006250993.1 | 9 | 1114 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gabra2| Gene Symbol: Gabra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135779.1 | NP_001129251.1 | 9 | 1050 | ||
| XM_006250931.2 | XP_006250993.1 | 9 | 1114 | |||