siRNA Details
| Gene Symbol: Ube2k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106006.1 | NP_001099476.1 | 3, 4 | 436 | ||
| XM_006250934.2 | XP_006250996.1 | 3, 4 | 633 | |||
| GenBank mRNA | FQ226880.1 | - | 3, 4 | 428 | ||
| FQ233155.1 | - | 3, 4 | 384 | |||
| FQ234076.1 | - | 3, 4 | 414 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_016786, AK048194, AK036068, BC002013, AK166647, and others.

Ube2k
Genomic Map