siRNA Details
| Gene Symbol: Pgm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106007.1 | NP_001099477.1 | 1, 2 | 153 | ||
| XM_006250996.2 | XP_006251058.1 | 1, 2 | 161 | |||
| GenBank mRNA | BC160893.1 | - | 1, 2 | 158 | 7445317 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pgm2| Gene Symbol: Pgm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106007.1 | NP_001099477.1 | 1, 2 | 153 | ||
| XM_006250996.2 | XP_006251058.1 | 1, 2 | 161 | |||
| GenBank mRNA | BC160893.1 | - | 1, 2 | 158 | 7445317 | |