siRNA Details
| Gene Symbol: Lap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011910.1 | NP_001011910.1 | 5 | 526 | ||
| GenBank mRNA | BC079381.1 | - | 5 | 526 | 7111053 | |
| FQ218842.1 | - | 1 | 44 | |||
| FQ235301.1 | - | 5 | 534 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lap3| Gene Symbol: Lap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011910.1 | NP_001011910.1 | 5 | 526 | ||
| GenBank mRNA | BC079381.1 | - | 5 | 526 | 7111053 | |
| FQ218842.1 | - | 1 | 44 | |||
| FQ235301.1 | - | 5 | 534 | |||