siRNA Details
| Gene Symbol: Lap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011910.1 | NP_001011910.1 | 8, 9 | 1023 | ||
| GenBank mRNA | BC079381.1 | - | 8, 9 | 1023 | 7111053 | |
| FQ218842.1 | - | 4, 5 | 541 | |||
| FQ235301.1 | - | 8, 9 | 1033 | |||

Lap3
Genomic Map