siRNA Details
| Gene Symbol: RGD1565560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_223504.4 | XP_223504.2 | 1 | 212 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1565560| Gene Symbol: RGD1565560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_223504.4 | XP_223504.2 | 1 | 212 | ||