siRNA Details
| Gene Symbol: Lyar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011911.1 | NP_001011911.1 | 8, 9 | 1184 | ||
| XM_006251087.1 | XP_006251149.1 | 8, 9 | 1190 | |||
| GenBank mRNA | BC079008.1 | - | 8, 9 | 1184 | 7113133 | |

Lyar
Genomic Map