siRNA Details
| Gene Symbol: Evc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170439.1 | NP_001163910.1 | 6 | 843 | ||
| XM_006251090.2 | XP_006251152.1 | 6 | 877 | |||
| XM_006251091.1 | XP_006251153.1 | 3 | 340 | |||
| GenBank mRNA | AB107664.1 | - | 3 | 393 | ||

Evc
Genomic Map